Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_015348.1 | 1 | 100 | |||
GenBank mRNA | U20366.1 | - | LIT1565 Hoxa11as |
1 | 100 | |
U20367.1 | - | LIT1564 Hoxa11as |
1 | 261 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ964700.1 | - | 1 | 319 | Chr. 6 - 52245052 - 52249256 [+] |