Gene Transcripts
- Gene Symbol
- Krit1
- Entrez Gene ID
- 79264
- Gene Name
- KRIT1, ankyrin repeat containing
- Gene Aliases
- 2010007K12Rik, A630036P20Rik, AA432855, AI450393, AI643869, BB155247, BB235701, Ccm1
- Chromosome Location
- Chr. 5: 3803165 - 3844515 on Build GRCm38
- UniGene ID
- Mm.32368
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033173.1 |
|
9 |
1520 |
|
GenBank mRNA |
BC044820.1 |
- |
8 |
1162 |
|
BC054819.1 |
- |
9 |
1512 |
|
View Full Product Details