Gene Transcripts

Gene Symbol
Etohd2
Entrez Gene ID
13996
Gene Name
ethanol decreased 2
Gene Aliases
AW491659
Chromosome Location
Chr. 13: 59769966 - 59773680 on Build GRCm38
UniGene ID
Mm.490435
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015349.2 2 1331
GenBank mRNA AK017511.1 - 2 1122
AK035969.1 - 2 1332
AK042838.1 - 2 1275

View Full Product Details