Gene Transcripts
- Gene Symbol
- Etohd2
- Entrez Gene ID
- 13996
- Gene Name
- ethanol decreased 2
- Gene Aliases
- AW491659
- Chromosome Location
- Chr. 13: 59769966 - 59773680 on Build GRCm38
- UniGene ID
- Mm.490435
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015349.2 |
|
2 |
2338 |
|
GenBank mRNA |
AK035969.1 |
- |
2 |
2339 |
|
View Full Product Details