Gene Transcripts
- Gene Symbol
- Snora16a
- Entrez Gene ID
- 100310813
- Gene Name
- small nucleolar RNA, H/ACA box 16A
- Gene Aliases
- Chromosome Location
- Chr. 4: 132309465 - 132309600 on Build GRCm38
- UniGene ID
- Mm.482271
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029412.1 |
|
1 |
106 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA977667.1 |
- |
1 |
224 |
|
Chr. 4 - 132309347 - 132310223 [+] |
View Full Product Details