Gene Transcripts
- Gene Symbol
- Snhg10
- Entrez Gene ID
- 69434
- Gene Name
- small nucleolar RNA host gene 10
- Gene Aliases
- 1700026B20Rik, AI642517
- Chromosome Location
- Chr. 12: 105030617 - 105032279 on Build GRCm38
- UniGene ID
- Mm.380736
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003145.3 |
|
2 |
476 |
|
GenBank mRNA |
AK006372.1 |
- |
2 |
477 |
|
BX524976.1 |
- |
2 |
437 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI154321.1 |
- |
2 |
434 |
|
Chr. 12 - 105030652 - 105032239 [-] |
GenBankmRNA |
BY706437.1 |
- |
2 |
474 |
|
Chr. 12 - 105030656 - 105032277 [-] |
GenBankmRNA |
AA981788.1 |
- |
2 |
444 |
|
Chr. 12 - 105030645 - 105032232 [-] |
View Full Product Details