Gene Transcripts
- Gene Symbol
- Pea15b
- Entrez Gene ID
- 231332
- Gene Name
- phosphoprotein enriched in astrocytes 15B
- Gene Aliases
- Chromosome Location
- Chr. 5: 77510039 - 77511003 on Build GRCm38
- UniGene ID
- Mm.373616
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC132661.1 |
- |
|
498 |
|
BC132663.1 |
- |
|
498 |
40130940 |
View Full Product Details