Gene Transcripts

Gene Symbol
NHEG1
Entrez Gene ID
100294720
Gene Name
neuroblastoma highly expressed 1
Gene Aliases
Chromosome Location
Chr. 6: 136982159 - 136993231 on Build GRCh38
UniGene ID
Hs.520412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027994.1 3 443
GenBank mRNA BC025319.1 - 3 446
BC033111.1 - 3 446
BC080649.1 - 3 446


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000432330.5 - 3 446 Chr. 6 - 136982165 - 136993234 [-]
GENCODE ENST00000418699.1 - 3 438 Chr. 6 - 136982165 - 136993234 [-]
GenBankmRNA BE260250.1 - 3 427 Chr. 6 - 136982925 - 136993215 [-]
GenBankmRNA BE312692.1 - 3 427 Chr. 6 - 136982870 - 136993215 [-]
GenBankmRNA BE262296.1 - 3 427 Chr. 6 - 136982849 - 136993215 [-]
GenBankmRNA BF206844.1 - 445 Chr. - - [-]
GenBankmRNA AL522654.3 - 3 443 Chr. 6 - 136982640 - 136993231 [-]
GenBankmRNA BE261212.1 - 3 428 Chr. 6 - 136982883 - 136993215 [-]

View Full Product Details