Gene Transcripts
- Gene Symbol
- MIR10B
- Entrez Gene ID
- 406903
- Gene Name
- microRNA 10b
- Gene Aliases
- MIRN10B, hsa-mir-10b, miRNA10B, mir-10b
- Chromosome Location
- Chr. 2: 176150303 - 176150412 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029609.1 |
|
1 |
44 |
|
View Full Product Details