Gene Transcripts
- Gene Symbol
- DIRC3
- Entrez Gene ID
- 729582
- Gene Name
- disrupted in renal carcinoma 3
- Gene Aliases
- Chromosome Location
- Chr. 2: 217284023 - 217756593 on Build GRCh38
- UniGene ID
- Hs.572788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026597.1 |
|
13 |
3597 |
|
GenBank mRNA |
AK024261.1 |
- |
12 |
2764 |
|
View Full Product Details