Gene Transcripts

Gene Symbol
CLLU1
Entrez Gene ID
574028
Gene Name
chronic lymphocytic leukemia up-regulated 1
Gene Aliases
Chromosome Location
Chr. 12: 92421531 - 92431002 on Build GRCh38
UniGene ID
Hs.730377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025233.2 NP_001020404.1 2 3931
NR_027932.1 3 2817
NR_027933.1 2 2260
GenBank mRNA AJ845162.1 - 2 2260

View Full Product Details