Gene Transcripts
- Gene Symbol
- RRN3P1
- Entrez Gene ID
- 730092
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 21796630 - 21819174 on Build GRCh38
- UniGene ID
- Hs.31290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003370.2 |
|
6, 7 |
511 |
|
GenBank mRNA |
CN397262.1 |
- |
6, 7 |
518 |
|
- Gene Symbol
- RRN3
- Entrez Gene ID
- 54700
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor
- Gene Aliases
- A-270G1.2, TIFIA
- Chromosome Location
- Chr. 16: 15060022 - 15094335 on Build GRCh38
- UniGene ID
- Hs.460078
- Species
- Homo sapiens
View Full Product Details