Gene Transcripts
- Gene Symbol
- SNORD22
- Entrez Gene ID
- 9304
- Gene Name
- small nucleolar RNA, C/D box 22
- Gene Aliases
- RNU22, U22
- Chromosome Location
- Chr. 11: 62852910 - 62853035 on Build GRCh38
- UniGene ID
- Hs.744737
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_000008.2 |
|
1 |
98 |
|
- Gene Symbol
- SNHG1
- Entrez Gene ID
- 23642
- Gene Name
- small nucleolar RNA host gene 1
- Gene Aliases
- LINC00057, NCRNA00057, U22HG, UHG
- Chromosome Location
- Chr. 11: 62851988 - 62855888 on Build GRCh38
- UniGene ID
- Hs.655643
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095849.1 |
- |
4 |
1948 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN337007.1 |
- |
9 |
421 |
|
Chr. 11 - 62852749 - 62855460 [-] |
GenBankmRNA |
L36588.3 |
- |
1 |
893 |
|
Chr. 11 - 62851914 - 62853833 [-] |
GenBankmRNA |
CB125560.1 |
- |
9 |
529 |
|
Chr. 11 - 62852832 - 62855885 [-] |
GenBankmRNA |
BM548627.1 |
- |
|
523 |
|
Chr. - - [-] |
GenBankmRNA |
U40580.3 |
- |
1 |
3442 |
|
Chr. 11 - 62851914 - 62856379 [-] |
GenBankmRNA |
DA728354.1 |
- |
9 |
527 |
|
Chr. 11 - 62852592 - 62855883 [-] |
View Full Product Details