Gene Transcripts
- Gene Symbol
- DGCR9
- Entrez Gene ID
- 25787
- Gene Name
- DiGeorge syndrome critical region gene 9 (non-protein coding)
- Gene Aliases
- DGS-A
- Chromosome Location
- Chr. 22: 19017834 - 19020248 on Build GRCh38
- UniGene ID
- Hs.106311
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024159.1 |
|
1 |
1166 |
|
GenBank mRNA |
BC036821.1 |
- |
1 |
1177 |
|
L77571.1 |
- |
1 |
1064 |
|
View Full Product Details