Gene Transcripts
- Gene Symbol
- OSTCP1
- Entrez Gene ID
- 202459
- Gene Name
- oligosaccharyltransferase complex subunit pseudogene 1
- Gene Aliases
- DC2L, OSTCL
- Chromosome Location
- Chr. 6: 158841117 - 158857632 on Build GRCh38
- UniGene ID
- Hs.487035
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028496.1 |
|
3 |
926 |
|
GenBank mRNA |
BC024224.1 |
- |
3 |
930 |
|
View Full Product Details