Gene Transcripts
- Gene Symbol
- SCARNA27
- Entrez Gene ID
- 100124533
- Gene Name
- small Cajal body-specific RNA 27
- Gene Aliases
- Chromosome Location
- Chr. 6: 8086408 - 8086533 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003703.1 |
|
1 |
51 |
|
View Full Product Details