Gene Transcripts

Gene Symbol
TTTY12
Entrez Gene ID
83867
Gene Name
testis-specific transcript, Y-linked 12 (non-protein coding)
Gene Aliases
NCRNA00135, TTY11, TTY12
Chromosome Location
Chr. Y: 7804924 - 7810683 on Build GRCh38
UniGene ID
Hs.667616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001551.1 2 913


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF332241.3 - 2 913 Chr. Y - 7804924 - 7810683 [+]

View Full Product Details