Gene Transcripts
- Gene Symbol
- GUSBP4
- Entrez Gene ID
- 375513
- Gene Name
- glucuronidase, beta pseudogene 4
- Gene Aliases
- C6orf216, GUSBL2, SMA3-L, SMAC3L
- Chromosome Location
- Chr. 6: 57908553 - 57936929 on Build GRCh38
- UniGene ID
- Hs.561539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132999.1 |
|
5 |
1206 |
|
GenBank mRNA |
AL831827.1 |
- |
1 |
1584 |
|
- Gene Symbol
- GUSBP2
- Entrez Gene ID
- 387036
- Gene Name
- glucuronidase, beta pseudogene 2
- Gene Aliases
- GUSBL1, GUSBP4, SMA3-L, SMAC3L, SMAC3L2, b55C20.1, bA239L20.1, bA239L20.5, bGLU-Lp
- Chromosome Location
- Chr. 6: 26871487 - 26956554 on Build GRCh38
- UniGene ID
- Hs.675815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003504.3 |
|
9 |
1144 |
|
GenBank mRNA |
BC034017.1 |
- |
9 |
1145 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB104995.1 |
- |
1 |
25 |
|
Chr. 5 - 20700419 - 20700517 [+] |
GenBankmRNA |
BX094662.1 |
- |
1 |
520 |
|
Chr. 5 - 34168239 - 34168957 [-] |
GenBankmRNA |
CB104809.1 |
- |
1 |
25 |
|
Chr. 5 - 20700419 - 20700517 [+] |
View Full Product Details