Gene Transcripts

Gene Symbol
FAM95B1
Entrez Gene ID
100133036
Gene Name
family with sequence similarity 95 member B1
Gene Aliases
Chromosome Location
Chr. 9: 40323571 - 40329220 on Build GRCh38
UniGene ID
Hs.584289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026759.1 3 940
GenBank mRNA AL833349.1 - 1 532

Gene Symbol
LOC105379252
Entrez Gene ID
105379252
Gene Name
family with sequence similarity 95 member B1-like
Gene Aliases
Chromosome Location
Chr. 9: 66047085 - 66052734 on Build GRCh38
UniGene ID
Hs.584289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136310.1 3 940

View Full Product Details