Gene Transcripts
- Gene Symbol
- FOXO3B
- Entrez Gene ID
- 2310
- Gene Name
- forkhead box O3B pseudogene
- Gene Aliases
- FKHRL1P1, ZNF286C
- Chromosome Location
- Chr. 17: 18665923 - 18673181 on Build GRCh38
- UniGene ID
- Hs.592126
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026718.1 |
|
1 |
3453 |
|
View Full Product Details