Gene Transcripts

Gene Symbol
BBIP1
Entrez Gene ID
92482
Gene Name
BBSome interacting protein 1
Gene Aliases
BBIP10, BBS18, NCRNA00081, bA348N5.3
Chromosome Location
Chr. 10: 110898730 - 110919366 on Build GRCh38
UniGene ID
Hs.232543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195304.1 NP_001182233.1 5 849
NM_001195305.1 NP_001182234.1 4 692
NM_001195306.1 NP_001182235.1 4 538
NM_001195307.1 NP_001182236.1 3 617
NM_001243783.1 NP_001230712.1 3 599
GenBank mRNA AK025724.1 - 4 512
AK125829.1 - 1 4696
AK307126.1 - 4 538
AK307314.1 - 5 725
BC105804.1 - 5 688
BI552279.1 - 3 431


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA638617.1 - 4 540 Chr. 10 - 110900214 - 110918936 [-]
GenBankmRNA DA091276.1 - 4 475 Chr. 10 - 110900145 - 110919149 [-]
GenBankmRNA BU786346.1 - 4 467 Chr. 10 - 110900196 - 110919141 [-]
GenBankmRNA BG204749.1 - 2 357 Chr. 10 - 110899891 - 110901612 [-]
GenBankmRNA DA090050.1 - 4 512 Chr. 10 - 110900204 - 110919186 [-]
GenBankmRNA CD642448.1 - 3 458 Chr. 10 - 110899908 - 110918213 [-]
GenBankmRNA AV706096.2 - 4 492 Chr. 10 - 110900220 - 110919166 [-]
GenBankmRNA DA712428.1 - 4 496 Chr. 10 - 110900186 - 110919169 [-]
GenBankmRNA DB170729.1 - 4 506 Chr. 10 - 110900196 - 110919179 [-]
GenBankmRNA DA179415.1 - 4 478 Chr. 10 - 110900174 - 110919152 [-]
GenBankmRNA DA973662.1 - 4 506 Chr. 10 - 110900185 - 110919180 [-]
GenBankmRNA DA346100.1 - 4 535 Chr. 10 - 110900194 - 110918933 [-]
GenBankmRNA BU189144.1 - 392 Chr. - - [-]
GenBankmRNA CB996772.1 - 4 526 Chr. 10 - 110899984 - 110919173 [-]
GenBankmRNA DA562097.1 - 4 517 Chr. 10 - 110900194 - 110919191 [-]
GenBankmRNA DB176723.1 - 4 506 Chr. 10 - 110900204 - 110919180 [-]
GenBankmRNA BU569868.1 - 4 478 Chr. 10 - 110900003 - 110919149 [-]
GenBankmRNA AW964060.1 - 4 466 Chr. 10 - 110900110 - 110919140 [-]
GenBankmRNA BG501418.1 - 5 612 Chr. 10 - 110900238 - 110919182 [-]
GenBankmRNA BQ213912.1 - 4 600 Chr. 10 - 110900003 - 110919274 [-]
GenBankmRNA BG427761.1 - 4 487 Chr. 10 - 110900057 - 110919157 [-]
GenBankmRNA DB557413.2 - 4 518 Chr. 10 - 110900150 - 110919187 [-]
GenBankmRNA DA646252.1 - 4 515 Chr. 10 - 110899960 - 110919188 [-]
GenBankmRNA DC340416.1 - 3 490 Chr. 10 - 110900174 - 110907816 [-]
GenBankmRNA DB267028.1 - 4 493 Chr. 10 - 110900197 - 110919167 [-]
GenBankmRNA AW173611.1 - 2 316 Chr. 10 - 110900059 - 110901575 [-]
GenBankmRNA BF967368.1 - 4 564 Chr. 10 - 110900033 - 110919232 [-]
GenBankmRNA DA274746.1 - 4 478 Chr. 10 - 110900157 - 110919152 [-]
GenBankmRNA DA826019.1 - 4 475 Chr. 10 - 110900174 - 110919149 [-]
GenBankmRNA AA210759.1 - 3 440 Chr. 10 - 110900215 - 110918197 [-]
GenBankmRNA DA287524.1 - 3 420 Chr. 10 - 110900119 - 110919187 [-]
GenBankmRNA DA520878.1 - 3 444 Chr. 10 - 110900129 - 110919193 [-]
GenBankmRNA CD356707.1 - 2 359 Chr. 10 - 110900092 - 110901614 [-]
GenBankmRNA BG708297.1 - 4 485 Chr. 10 - 110900021 - 110919156 [-]
GenBankmRNA BX426107.2 - 482 Chr. - - [-]
GenBankmRNA BG195536.1 - 2 354 Chr. 10 - 110900227 - 110901612 [-]
GenBankmRNA DA176600.1 - 4 538 Chr. 10 - 110900218 - 110918936 [-]
GenBankmRNA DA480252.1 - 4 499 Chr. 10 - 110900186 - 110919173 [-]
GenBankmRNA DC393891.1 - 4 544 Chr. 10 - 110900217 - 110919152 [-]
GenBankmRNA DA012980.1 - 4 493 Chr. 10 - 110900175 - 110919167 [-]
GenBankmRNA BF244359.1 - 3 416 Chr. 10 - 110900066 - 110919182 [-]
GenBankmRNA DA365239.1 - 4 491 Chr. 10 - 110900170 - 110919164 [-]
GenBankmRNA DB059835.1 - 4 512 Chr. 10 - 110900220 - 110919186 [-]
GenBankmRNA CX872108.1 - 3 451 Chr. 10 - 110899848 - 110918213 [-]
GenBankmRNA DA054554.1 - 3 425 Chr. 10 - 110900104 - 110919191 [-]
GenBankmRNA DA385629.1 - 4 517 Chr. 10 - 110900182 - 110919191 [-]
GenBankmRNA BG699263.1 - 4 486 Chr. 10 - 110900007 - 110919158 [-]
GenBankmRNA DB109839.1 - 4 519 Chr. 10 - 110900193 - 110919192 [-]
GenBankmRNA DA070805.1 - 3 426 Chr. 10 - 110900109 - 110919193 [-]
GenBankmRNA DB028886.1 - 4 500 Chr. 10 - 110900197 - 110919174 [-]
GenBankmRNA BE867726.1 - 3 454 Chr. 10 - 110899987 - 110918213 [-]
GenBankmRNA BU852778.1 - 4 474 Chr. 10 - 110900115 - 110919145 [-]
GenBankmRNA BG655027.1 - 3 389 Chr. 10 - 110900175 - 110918155 [-]
GenBankmRNA BU561307.1 - 5 654 Chr. 10 - 110900176 - 110919167 [-]
GenBankmRNA DA280162.1 - 4 493 Chr. 10 - 110900181 - 110919167 [-]
GenBankmRNA BI254459.1 - 4 475 Chr. 10 - 110900151 - 110919149 [-]
GenBankmRNA CF994690.1 - 4 546 Chr. 10 - 110900133 - 110919195 [-]
GenBankmRNA AW968916.1 - 3 446 Chr. 10 - 110899943 - 110918213 [-]
GenBankmRNA AV761413.2 - 3 396 Chr. 10 - 110900214 - 110919163 [-]
GenBankmRNA AA398149.1 - 3 481 Chr. 10 - 110900236 - 110907807 [-]
GenBankmRNA DA171687.1 - 4 475 Chr. 10 - 110900145 - 110919149 [-]
GenBankmRNA DA017962.1 - 4 483 Chr. 10 - 110900155 - 110919157 [-]
GenBankmRNA BX441766.2 - 4 482 Chr. 10 - 110899867 - 110919156 [-]
GenBankmRNA CR737308.1 - 3 447 Chr. 10 - 110899960 - 110918213 [-]
GenBankmRNA DA483836.1 - 4 478 Chr. 10 - 110900202 - 110919152 [-]
GenBankmRNA CD238043.1 - 4 479 Chr. 10 - 110900141 - 110919153 [-]
GenBankmRNA DB516410.1 - 3 445 Chr. 10 - 110900234 - 110918212 [-]
GenBankmRNA DA657827.1 - 4 511 Chr. 10 - 110900185 - 110919185 [-]
GenBankmRNA DA683617.1 - 4 478 Chr. 10 - 110900234 - 110919152 [-]
GenBankmRNA DA364841.1 - 4 517 Chr. 10 - 110900208 - 110919191 [-]
GenBankmRNA CX872684.1 - 4 493 Chr. 10 - 110899928 - 110919166 [-]
GenBankmRNA DB211776.1 - 3 431 Chr. 10 - 110900134 - 110919173 [-]
GenBankmRNA BM153163.1 - 3 437 Chr. 10 - 110900211 - 110919201 [-]
GenBankmRNA DA365363.1 - 4 513 Chr. 10 - 110900185 - 110919187 [-]
GenBankmRNA DB507525.2 - 4 519 Chr. 10 - 110900234 - 110919193 [-]
GenBankmRNA DA076311.1 - 4 482 Chr. 10 - 110900166 - 110919154 [-]
GenBankmRNA DB442650.2 - 4 519 Chr. 10 - 110900233 - 110919193 [-]
GenBankmRNA CN388948.1 - 4 491 Chr. 10 - 110900165 - 110919165 [-]
GenBankmRNA DA258827.1 - 4 499 Chr. 10 - 110900188 - 110919173 [-]
GenBankmRNA DA346930.1 - 4 483 Chr. 10 - 110900171 - 110919157 [-]
GenBankmRNA DB487968.2 - 4 519 Chr. 10 - 110900150 - 110919193 [-]
GenBankmRNA DA694218.1 - 4 513 Chr. 10 - 110900208 - 110919187 [-]
GenBankmRNA AV753498.1 - 3 451 Chr. 10 - 110900047 - 110918213 [-]

View Full Product Details