Gene Transcripts
- Gene Symbol
- ANKRD30BP2
- Entrez Gene ID
- 149992
- Gene Name
- ankyrin repeat domain 30B pseudogene 2
- Gene Aliases
- C21orf99, CT85, CTSP-1, CTSP1
- Chromosome Location
- Chr. 21: 13038166 - 13118251 on Build GRCh38
- UniGene ID
- Hs.558645
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026916.1 |
|
12 |
3230 |
|
GenBank mRNA |
DQ179141.1 |
- |
3 |
445 |
|
View Full Product Details