Gene Transcripts
- Gene Symbol
- DSCR8
- Entrez Gene ID
- 84677
- Gene Name
- Down syndrome critical region 8
- Gene Aliases
- C21orf65, CT25.1a, CT25.1b, MMA-1, MMA-1a, MMA-1b, MMA1, MTAG2
- Chromosome Location
- Chr. 21: 38121451 - 38156511 on Build GRCh38
- UniGene ID
- Hs.733345
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026838.1 |
|
1, 2 |
162 |
|
NR_026839.1 |
|
1, 2 |
162 |
|
NR_026840.1 |
|
1, 2 |
162 |
|
NR_026841.1 |
|
1, 2 |
162 |
|
NR_026842.1 |
|
1, 2 |
162 |
|
GenBank mRNA |
AF321193.1 |
- |
1, 2 |
137 |
|
AF426256.1 |
- |
1, 2 |
85 |
|
AJ001904.1 |
- |
1, 2 |
122 |
|
AJ297914.2 |
- |
1, 2 |
140 |
|
AJ301615.2 |
- |
1, 2 |
140 |
|
AJ305085.2 |
- |
1, 2 |
140 |
|
AJ306839.2 |
- |
1, 2 |
140 |
|
AJ306840.2 |
- |
1, 2 |
140 |
|
AJ783421.1 |
- |
1, 2 |
140 |
|
BC015981.1 |
- |
1, 2 |
144 |
|
BC029353.1 |
- |
1, 2 |
162 |
|
BC070229.1 |
- |
1, 2 |
121 |
|
BC117304.1 |
- |
1, 2 |
94 |
|
BC117330.1 |
- |
1, 2 |
94 |
|
BC143995.1 |
- |
1, 2 |
94 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF594279.1 |
- |
1, 2 |
149 |
|
Chr. 21 - 38121492 - 38156511 [+] |
GenBankmRNA |
BX118383.1 |
- |
1, 2 |
84 |
|
Chr. 21 - 38121529 - 38156506 [+] |
GenBankmRNA |
BG687098.1 |
- |
|
122 |
|
Chr. - - [-] |
View Full Product Details