Gene Transcripts

Gene Symbol
CROCCP2
Entrez Gene ID
84809
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 2
Gene Aliases
CROCCL1
Chromosome Location
Chr. 1: 16618256 - 16630906 on Build GRCh38
UniGene ID
Hs.631865
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026752.1 7 1345
GenBank mRNA AK128484.1 - 1656
BC006312.2 - 7 1599
BC010508.1 - 4 870
BC047736.1 - 7 1345
DQ786196.1 - 2 758

View Full Product Details