Gene Transcripts
- Gene Symbol
- KRT16P3
- Entrez Gene ID
- 644945
- Gene Name
- keratin 16 pseudogene 3
- Gene Aliases
- KERSMCR
- Chromosome Location
- Chr. 17: 20501513 - 20504498 on Build GRCh38
- UniGene ID
- Hs.572477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029393.1 |
|
7 |
1921 |
|
GenBank mRNA |
BC110641.1 |
- |
7 |
1904 |
|
- Gene Symbol
- KRT16P1
- Entrez Gene ID
- 729252
- Gene Name
- keratin 16 pseudogene 1
- Gene Aliases
- KRT14P, KRT16P
- Chromosome Location
- Chr. 17: 18439908 - 18442895 on Build GRCh38
- UniGene ID
- Hs.585811
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073414.1 |
|
7 |
1781 |
|
View Full Product Details