Gene Transcripts
- Gene Symbol
- UGT3A2
- Entrez Gene ID
- 167127
- Gene Name
- UDP glycosyltransferase family 3 member A2
- Gene Aliases
- Chromosome Location
- Chr. 5: 36035017 - 36071358 on Build GRCh38
- UniGene ID
- Hs.348941
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_031764.1 |
|
3, 4 |
439 |
|
GenBank mRNA |
AK303154.1 |
- |
3, 4 |
400 |
|
View Full Product Details