Gene Transcripts

Gene Symbol
CXCR2P1
Entrez Gene ID
3580
Gene Name
C-X-C motif chemokine receptor 2 pseudogene 1
Gene Aliases
CXCR2P, IL8RBP
Chromosome Location
Chr. 2: 218059155 - 218061290 on Build GRCh38
UniGene ID
Hs.647858
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002712.1
1 265
GenBank mRNA BC033255.1 - LIT2061
IL8RBP
1 265


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM008326.1 - 1 266 Chr. 2 - 218060405 - 218061291 [-]

View Full Product Details