Gene Transcripts
- Gene Symbol
- MTMR9LP
- Entrez Gene ID
- 339483
- Gene Name
- myotubularin related protein 9-like, pseudogene
- Gene Aliases
- MTMR9L
- Chromosome Location
- Chr. 1: 32231660 - 32241710 on Build GRCh38
- UniGene ID
- Hs.471067
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB549336.1 |
- |
2 |
401 |
|
Chr. 1 - 32231661 - 32232184 [-] |
GenBankmRNA |
BM833152.1 |
- |
4 |
538 |
|
Chr. 1 - 32231698 - 32234981 [-] |
View Full Product Details