Gene Transcripts

Gene Symbol
SNORA24
Entrez Gene ID
677809
Gene Name
small nucleolar RNA, H/ACA box 24
Gene Aliases
ACA24, SNORA24A
Chromosome Location
Chr. 4: 118279190 - 118279320 on Build GRCh38
UniGene ID
Hs.744742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002963.1 1 65
GenBank mRNA AJ609460.1 - 1 65
AJ609478.1 - 1 65


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG538966.1 - 1 498 Chr. 4 - 118278758 - 118279476 [+]
GenBankmRNA BQ722308.1 - 1 477 Chr. 4 - 118278778 - 118279554 [+]

View Full Product Details