Gene Transcripts

Gene Symbol
TPRXL
Entrez Gene ID
348825
Gene Name
tetrapeptide repeat homeobox like
Gene Aliases
TPRX3P
Chromosome Location
Chr. 3: 13937307 - 14065984 on Build GRCh38
UniGene ID
Hs.529180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002223.3 3 1928
GenBank mRNA AK092426.1 - 3 1853
BC027729.2 - 5 1933


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA R82240.1 - 1 277 Chr. 3 - 14065373 - 14065785 [+]
GenBankmRNA BM764893.1 - 1 459 Chr. 3 - 14065191 - 14065879 [+]
GenBankmRNA BM811485.1 - 231 Chr. - - [-]
GenBankmRNA BM764898.1 - 1 459 Chr. 3 - 14065191 - 14065781 [+]
GenBankmRNA BX111860.1 - 1 277 Chr. 3 - 14065373 - 14065986 [+]

View Full Product Details