Gene Transcripts
- Gene Symbol
- PMCHL2
- Entrez Gene ID
- 5370
- Gene Name
- pro-melanin concentrating hormone like 2 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 5: 71375785 - 71385993 on Build GRCh38
- UniGene ID
- Hs.742169
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003922.1 |
|
2 |
1434 |
|
GenBank mRNA |
AY008412.1 |
- |
2 |
1434 |
|
AY008413.1 |
- |
2 |
1434 |
|
- Gene Symbol
- PMCHL1
- Entrez Gene ID
- 5369
- Gene Name
- pro-melanin concentrating hormone like 1 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 5: 22142352 - 22152272 on Build GRCh38
- UniGene ID
- Hs.247975
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY008411.1 |
- |
2 |
1436 |
|
View Full Product Details