Gene Transcripts
- Gene Symbol
- ST13P4
- Entrez Gene ID
- 145165
- Gene Name
- suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 4
- Gene Aliases
- FAM10A4, FAM10A4P
- Chromosome Location
- Chr. 13: 50172018 - 50173616 on Build GRCh38
- UniGene ID
- Hs.511834
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002183.1 |
|
1 |
690 |
|
GenBank mRNA |
AF539468.1 |
- |
1 |
690 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF279660.3 |
- |
6 |
175665 |
|
Chr. 13 - 49996975 - 50344526 [+] |
View Full Product Details