Gene Transcripts
- Gene Symbol
- CSNK1A1P1
- Entrez Gene ID
- 161635
- Gene Name
- casein kinase 1 alpha 1 pseudogene 1
- Gene Aliases
- CSNK1A1P
- Chromosome Location
- Chr. 15: 36799100 - 36818506 on Build GRCh38
- UniGene ID
- Hs.659822
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027320.1 |
|
4 |
1014 |
|
GenBank mRNA |
BC028192.1 |
- |
4 |
967 |
|
BC066972.1 |
- |
4 |
805 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB527361.1 |
- |
4 |
312 |
|
Chr. 15 - 36799613 - 36817804 [-] |
GenBankmRNA |
BI825331.1 |
- |
4 |
603 |
|
Chr. 15 - 36799633 - 36818095 [-] |
GenBankmRNA |
DB518216.1 |
- |
4 |
267 |
|
Chr. 15 - 36799618 - 36817759 [-] |
View Full Product Details