Gene Transcripts
- Gene Symbol
- SCARNA1
- Entrez Gene ID
- 677774
- Gene Name
- small Cajal body-specific RNA 1
- Gene Aliases
- ACA35
- Chromosome Location
- Chr. 1: 27834401 - 27834566 on Build GRCh38
- UniGene ID
- Hs.689694
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002997.1 |
|
1 |
126 |
|
GenBank mRNA |
AJ609484.1 |
- |
1 |
126 |
|
View Full Product Details