Gene Transcripts

Gene Symbol
WHAMMP2
Entrez Gene ID
440253
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 2
Gene Aliases
WHAMML2, WHDC1L2
Chromosome Location
Chr. 15: 28737583 - 28758362 on Build GRCh38
UniGene ID
Hs.558967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026589.1 9 3135
GenBank mRNA AK091254.1 - 4 1818

View Full Product Details