Gene Transcripts
- Gene Symbol
- RRN3P2
- Entrez Gene ID
- 653390
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 16: 29074842 - 29116718 on Build GRCh38
- UniGene ID
- Hs.720348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003369.2 |
|
15, 16 |
1719 |
|
GenBank mRNA |
AK225670.1 |
- |
15, 16 |
1569 |
|
AK298442.1 |
- |
13, 14 |
1460 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM686819.1 |
- |
1, 2 |
51 |
|
Chr. 16 - 29104122 - 29116545 [+] |
View Full Product Details