Gene Transcripts
- Gene Symbol
- HAR1B
- Entrez Gene ID
- 768097
- Gene Name
- highly accelerated region 1B (non-protein coding)
- Gene Aliases
- HAR1R, LINC00065, NCRNA00065
- Chromosome Location
- Chr. 20: 63095493 - 63102319 on Build GRCh38
- UniGene ID
- Hs.635159
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003245.1 |
|
|
2 |
454 |
|
GenBank mRNA |
DQ860410.1 |
- |
LIT3338 HAR1R |
2 |
454 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY011775.1 |
- |
2 |
444 |
|
Chr. 20 - 63095871 - 63102309 [-] |
GenBankmRNA |
HY023235.1 |
- |
2 |
418 |
|
Chr. 20 - 63095892 - 63102283 [-] |
View Full Product Details