Gene Transcripts

Gene Symbol
CLEC4GP1
Entrez Gene ID
440508
Gene Name
C-type lectin domain family 4 member G pseudogene 1
Gene Aliases
Chromosome Location
Chr. 19: 7787484 - 7791012 on Build GRCh38
UniGene ID
Hs.568222
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002931.2 7 1878
GenBank mRNA AK090689.1 - 7 1860
AK090734.1 - 7 1862
AK123140.1 - 7 1844
BC047593.1 - 5 1752

View Full Product Details