Gene Transcripts

Gene Symbol
ASMTL-AS1
Entrez Gene ID
80161
Gene Name
ASMTL antisense RNA 1
Gene Aliases
ASMTL-AS, ASMTLAS, CXYorf2, NCRNA00105
Chromosome Location
Chr. Y: 1400531 - 1415421 on Build GRCh38
UniGene ID
Hs.521856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026710.1 2 715
NR_026711.1 2 715
GenBank mRNA AF193043.2 - 1 78
AK023392.1 - 2 148
BC093994.2 - 1 58
BC143527.1 - 1 58


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CT003675.1 - 1 310 Chr. X - 1401811 - 1413915 [+]
GenBankmRNA CF455161.1 - 227 Chr. - - [-]
GenBankmRNA BE830876.1 - 87 Chr. - - [-]

View Full Product Details