Gene Transcripts

Gene Symbol
MESTIT1
Entrez Gene ID
317751
Gene Name
MEST intronic transcript 1, antisense RNA
Gene Aliases
MEST-AS1, MEST-IT, MEST-IT1, NCRNA00040, PEG1-AS
Chromosome Location
Chr. 7: 130487057 - 130491172 on Build GRCh38
UniGene ID
Hs.627349
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_004382.1 2 399


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AW161444.1 - 2 181 Chr. 7 - 130489632 - 130490954 [-]
GenBankmRNA DB099005.1 - 2 260 Chr. 7 - 130489574 - 130491033 [-]
GENCODE ENST00000610046.2 - 260 Chr. 7 - 130486042 - 130486183 [-]
GenBankmRNA DB055510.1 - 2 263 Chr. 7 - 130489585 - 130491033 [-]
GenBankmRNA AW163454.1 - 2 181 Chr. 7 - 130489582 - 130490954 [-]
GenBankmRNA AF482998.3 - 2 399 Chr. 7 - 130487057 - 130491172 [-]

View Full Product Details