Gene Transcripts
- Gene Symbol
- PPP1R2P9
- Entrez Gene ID
- 80316
- Gene Name
- protein phosphatase 1 regulatory inhibitor subunit 2 pseudogene 9
- Gene Aliases
- I-4
- Chromosome Location
- Chr. X: 42777366 - 42778235 on Build GRCh38
- UniGene ID
- Hs.127689
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002191.2 |
|
1 |
346 |
|
GenBank mRNA |
AB044137.1 |
- |
1 |
346 |
|
BC056673.1 |
- |
1 |
276 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI832410.1 |
- |
1 |
266 |
|
Chr. X - 42777377 - 42778153 [-] |
GenBankmRNA |
BX096729.1 |
- |
1 |
358 |
|
Chr. X - 42777528 - 42778247 [-] |
View Full Product Details