Gene Transcripts
- Gene Symbol
- SNAPC2
- Entrez Gene ID
- 6618
- Gene Name
- small nuclear RNA activating complex polypeptide 2
- Gene Aliases
- PTFDELTA, SNAP45
- Chromosome Location
- Chr. 19: 7920309 - 7923251 on Build GRCh38
- UniGene ID
- Hs.631860
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030717.1 |
|
1 |
58 |
|
GenBank mRNA |
BP234322.1 |
- |
1 |
57 |
|
BX647416.1 |
- |
1 |
25 |
|
View Full Product Details