Gene Transcripts
- Gene Symbol
- MIR17HG
- Entrez Gene ID
- 407975
- Gene Name
- miR-17-92a-1 cluster host gene
- Gene Aliases
- C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92
- Chromosome Location
- Chr. 13: 91347820 - 91354575 on Build GRCh38
- UniGene ID
- Hs.24115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027349.1 |
|
3, 4 |
293 |
|
NR_027350.1 |
|
2 |
4384 |
|
GenBank mRNA |
AK024240.1 |
- |
1 |
2107 |
|
- Gene Symbol
- MIR18A
- Entrez Gene ID
- 406953
- Gene Name
- microRNA 18a
- Gene Aliases
- MIR18, MIRN18, MIRN18A, hsa-mir-18, hsa-mir-18a, miR-18, miRNA18A, mir-18a
- Chromosome Location
- Chr. 13: 91350751 - 91350821 on Build GRCh38
- UniGene ID
- Hs.24115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB176708.1 |
- |
LIT3364 oncomir-1 |
2 |
4384 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG772153.1 |
- |
3, 4 |
298 |
|
Chr. 13 - 91347817 - 91354466 [+] |
View Full Product Details