Gene Transcripts

Gene Symbol
PCGEM1
Entrez Gene ID
64002
Gene Name
PCGEM1, prostate-specific transcript (non-protein coding)
Gene Aliases
LINC00071, NCRNA00071, PCAT9
Chromosome Location
Chr. 2: 192749845 - 192776899 on Build GRCh38
UniGene ID
Hs.546994
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002769.1
3 762
GenBank mRNA AF223389.1 - LIT1562
PCGEM1
3 762

View Full Product Details