Gene Transcripts
- Gene Symbol
- SPRR2C
- Entrez Gene ID
- 6702
- Gene Name
- small proline rich protein 2C (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 1: 153140118 - 153141493 on Build GRCh38
- UniGene ID
- Hs.2421
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003062.1 |
|
2 |
158 |
|
GenBank mRNA |
BC120937.1 |
- |
2 |
116 |
|
M21539.1 |
- |
2 |
158 |
|
View Full Product Details