Gene Transcripts
- Gene Symbol
- ADGRE4P
- Entrez Gene ID
- 326342
- Gene Name
- adhesion G protein-coupled receptor E4, pseudogene
- Gene Aliases
- EMR4, EMR4P, FIRE, GPR127, PGR16
- Chromosome Location
- Chr. 19: 6952500 - 6990846 on Build GRCh38
- UniGene ID
- Hs.688043
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024075.1 |
|
6 |
505 |
|
GenBank mRNA |
AF489700.1 |
- |
6 |
505 |
|
View Full Product Details