Gene Transcripts

Gene Symbol
TAZ
Entrez Gene ID
6901
Gene Name
tafazzin
Gene Aliases
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
Chromosome Location
Chr. X: 154411406 - 154421726 on Build GRCh38
UniGene ID
Hs.409911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024048.2 3 611
XM_006724841.3 XP_006724904.1 3 276
XM_006724842.3 XP_006724905.1 3 276
XM_017029764.1 XP_016885253.1 1 22
XM_017029765.1 XP_016885254.1 3 723
GenBank mRNA AL531401.3 - 3 613
BC005062.1 - 3 568
CA488175.1 - 2 101
KU178351.1 - 3 285

View Full Product Details