Gene Transcripts
- Gene Symbol
- TPTEP1
- Entrez Gene ID
- 387590
- Gene Name
- transmembrane phosphatase with tensin homology pseudogene 1
- Gene Aliases
- psiTPTE22
- Chromosome Location
- Chr. 22: 16601911 - 16648830 on Build GRCh38
- UniGene ID
- Hs.474116
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG333521.1 |
- |
3 |
569 |
|
Chr. 22 - 16601911 - 16614237 [+] |
GenBankmRNA |
DR004418.1 |
- |
3 |
578 |
|
Chr. 22 - 16601920 - 16614123 [+] |
GenBankmRNA |
DB222954.1 |
- |
3 |
531 |
|
Chr. 22 - 16601947 - 16614146 [+] |
GenBankmRNA |
AW572313.1 |
- |
3 |
456 |
|
Chr. 22 - 16602021 - 16614178 [+] |
View Full Product Details