Gene Transcripts
- Gene Symbol
- AMZ2P1
- Entrez Gene ID
- 201283
- Gene Name
- archaelysin family metallopeptidase 2 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 17: 64966550 - 64975585 on Build GRCh38
- UniGene ID
- Hs.396447
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026903.1 |
|
1 |
952 |
|
GenBank mRNA |
AK056627.1 |
- |
1 |
935 |
|
BX537538.1 |
- |
1 |
826 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
T89821.1 |
- |
|
20 |
|
Chr. - - [-] |
GenBankmRNA |
BU533175.1 |
- |
|
191 |
|
Chr. - - [-] |
GenBankmRNA |
CD518505.1 |
- |
|
258 |
|
Chr. - - [-] |
GenBankmRNA |
AL537435.3 |
- |
1, 2 |
602 |
|
Chr. 17 - 64973435 - 64975545 [-] |
View Full Product Details