Gene Transcripts
- Gene Symbol
- TMED10P1
- Entrez Gene ID
- 286102
- Gene Name
- transmembrane p24 trafficking protein 10 pseudogene 1
- Gene Aliases
- TMED10P, Tmp21-II
- Chromosome Location
- Chr. 8: 144994865 - 144998897 on Build GRCh38
- UniGene ID
- Hs.744580
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002807.4 |
|
1 |
2255 |
|
View Full Product Details